Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyl Phosphite (contains ca. 5% Phenol) 75.0+%, TCI America™
SDP

Catalog No. D0907100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0907100G 100 g
D090725G 25 g
D0907500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0907100G Supplier TCI America Supplier No. D0907100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4712-55-4
Molecular Formula C12H10O3P+
Molecular Weight (g/mol) 233.183
MDL Number MFCD00044497
InChI Key CDXVUROVRIFQMV-UHFFFAOYSA-N
Synonym diphenyl phosphite, diphenyl phosphonate, phosphonic acid, diphenyl ester, diphenoxyphosphine oxide, diphenyl hydrogen phosphite, phosphorous acid, diphenyl ester, unii-5144js6xum, phosphonic acid diphenyl ester, phenyl phosphonate, pho 2hpo, phenyl phosphonate pho 2hpo
PubChem CID 6327546
IUPAC Name oxo(diphenoxy)phosphanium
SMILES C1=CC=C(C=C1)O[P+](=O)OC2=CC=CC=C2

Specifications

Melting Point 12°C
Color Yellow
Boiling Point 148°C
Quantity 100 g
UN Number 1760
Formula Weight 234.19
Percent Purity ≥75.0% (HPLC)
Physical Form Liquid
Chemical Name or Material Diphenyl Phosphite (contains ca. 5% Phenol)
EINECSNumber (3)-2602
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.