Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylacetic Acid 98.0+%, TCI America™
SDP

Catalog No. D0869100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0869100G 100 g
D086925G 25 g
D0869500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0869100G Supplier TCI America Supplier No. D0869100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 117-34-0
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.248
MDL Number MFCD00004251
InChI Key PYHXGXCGESYPCW-UHFFFAOYSA-N
Synonym diphenylacetic acid, diphenylethanoic acid, acetic acid, diphenyl, benzeneacetic acid, .alpha.-phenyl, 1,1-diphenylacetic acid, unii-658ncz0nko, diphenyl-acetic acid, alpha-toluic acid, alpha-phenyl, 658ncz0nko, benzeneacetic acid, alpha-phenyl
PubChem CID 8333
ChEBI CHEBI:41967
IUPAC Name 2,2-diphenylacetic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)O

Specifications

Melting Point 146°C
Color White
Quantity 100 g
Formula Weight 212.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenylacetic Acid
EINECSNumber (9)-1874
RTECSNumber AH2515000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.