Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylamine, 99%, pure

Catalog No. AC150740010
Change view
Click to view available options
Quantity:
5 g
100 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC150740010 1 kg Glass bottle
AC150740050 5 g Glass bottle
AC150741000 100 g Glass bottle
AC150740051 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC150740010 Supplier Thermo Scientific Chemicals Supplier No. 150740010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 122-39-4 | C12H11N | 169.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-39-4
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
InChI Key DMBHHRLKUKUOEG-UHFFFAOYSA-N
Synonym diphenylamine, n-phenylbenzenamine, anilinobenzene, benzenamine, n-phenyl, n,n-diphenylamine, phenylaniline, scaldip, n-phenylbenzeneamine, big dipper, no-scald
PubChem CID 11487
ChEBI CHEBI:4640
IUPAC Name N-phenylaniline
SMILES C1=CC=C(C=C1)NC2=CC=CC=C2

Specifications

Melting Point 52.0°C to 54.0°C
Color Orange to White
Boiling Point 302.0°C
Flash Point 152°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)2NH
Quantity 1 kg
Beilstein 12,174
Merck Index 15,3347
Solubility Information Solubility in water: 0.05g/L. Other solubilities: soluble in alcohol and propyl alcohol, freely soluble in benzene, ether, glacial acetic, acid and carbon disulfide
Formula Weight 169.23
Percent Purity 99%
Grade Pure
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Diphenylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 204-539-4
Recommended Storage This reagent discolors on exposure to light

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.