Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylamine Sulfate 98.0+%, TCI America™
SDP

Catalog No. D087525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D087525G 25 g
D0875500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D087525G Supplier TCI America Supplier No. D087525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 587-84-8
Molecular Formula C12H13NO4S
Molecular Weight (g/mol) 267.30
MDL Number MFCD00042016
InChI Key IPZMDJYHJNHGML-UHFFFAOYSA-N
Synonym diphenylamine sulfate, benzenamine, n-phenyl-, sulfate 1:1, usaf ek-743, n-phenylaniline; sulfuric acid, diphenylamine, hydrogen sulfate, diphenylammonium hydrogen sulphate, diphenylamine; sulfuric acid, diphenylaminesulfate, bis n-phenylanilinium sulphate, diphenylaminsulfat
PubChem CID 11486
IUPAC Name N-phenylaniline; sulfuric acid
SMILES OS(O)(=O)=O.N(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 150°C
Color White
Quantity 25 g
Formula Weight 267.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenylamine Sulfate
RTECSNumber JJ9285000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.