Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyldithiophosphonic acid

Catalog No. AA1775804
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1775804 2 g
AA1775802 0.5 g
AA1775809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1775804 Supplier Thermo Scientific Chemicals Supplier No. 01775804
Only null left
Add to Cart

CAS: 1015-38-9 | C12H11PS2 | 250.31 g/mol

Diphenyldithiophosphonic acid is used in place of phenyl phosphate for acylation of nucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1015-38-9
Molecular Formula C12H11PS2
Molecular Weight (g/mol) 250.31
MDL Number MFCD00014457
InChI Key CLUOCCWZZAGLPM-UHFFFAOYSA-N
Synonym diphenylphosphinodithioic acid, diphenyldithiophosphonic acid, diphenyldithiophosphinic acid, phosphinodithioic acid, diphenyl, diphenyl sulfanyl-??-phosphanethione, dithiodiphenylphosphinic acid, diphenyl-sulfanyl-sulfanylidene, diphenyl sulfanyl-lambda-phosphanethione, diphenyl-sulfanyl-sulfanylidenephosphorane, diphenyl sulfanyl-$l; 5-phosphanethione
PubChem CID 129564
IUPAC Name diphenyl-sulfanyl-sulfanylidene-$l^{5}-phosphane
SMILES SP(=S)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 55°C to 56°C
Quantity 2 g
Formula Weight 250.31
Physical Form Crystalline
Chemical Name or Material Diphenyldithiophosphonic acid
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.