Learn More
Enzo Life Sciences Diphenyleneiodonium chloride (10mg). CAS: 4673-26-1
SDP
Supplier:  Enzo Life Sciences BMLCN2400010
Encompass_Preferred
Binds strongly to flavoproteins and is thus a powerful and specific inhibitor of several important enzymes, including nitric oxide synthase (NOS), NADPH-ubiquinone oxidoreductase, NADPH oxidases and NADPH cytochrome P450 oxidoreductase. Nitric oxide synthase, which shows significant homology with cytochrome P450 reductase, has been shown to be irreversibly inhibited by this compound in cultured peritoneal macrophages (IC50=30nM). Alternative name: DPI chloride, Dibenziodolium chloride. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (warm) (5mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0457
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.