Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylgermanium dichloride, 98+%

Catalog No. AA3411306
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3411306 5 g
AA3411303 1 g
AA3411314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3411306 Supplier Thermo Scientific Chemicals Supplier No. 03411306
Add to Cart

CAS: 1613-66-7 | C12H14Cl2Ge | 301.77 g/mol

Diphenylgermanium dichloride is used as an organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1613-66-7
Molecular Formula C12H14Cl2Ge
Molecular Weight (g/mol) 301.77
MDL Number MFCD00013587
InChI Key YROQISGFJISKEN-UHFFFAOYSA-N
Synonym diphenyldichlorogermane, diphenylgermanium dichloride, germane, dichlorodiphenyl, dichlorodiphenylgermene, dichloro diphenyl germane, acmc-1bssq, germane,dichlorodiphenyl, germanium diphenyl dichloride
PubChem CID 74167
IUPAC Name dichloro(diphenyl)germane
SMILES Cl.Cl.[GeH2](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 9°C
Density 1.415 g/mL
Boiling Point 107°C (16 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.597
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 297.73
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Diphenylgermanium dichloride
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 216-562-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.