Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals alpha,alpha-Diphenylglycine, 98%

Catalog No. AC171720250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC171720250 25 g Glass bottle
AC171720050 5 g Glass bottle
AC171721000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC171720250 Supplier Thermo Scientific Chemicals Supplier No. 171720250
Add to Cart

CAS: 3060-50-2 | C14H13NO2 | 227.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3060-50-2
Molecular Formula C14H13NO2
Molecular Weight (g/mol) 227.26
MDL Number MFCD00008048
InChI Key YBONNYNNFBAKLI-UHFFFAOYSA-N
Synonym 2,2-diphenylglycine, alpha,alpha-diphenylglycine, alpha-aminodiphenylacetic acid, glycine, 2,2-diphenyl, alpha-amino-alpha-phenylbenzeneacetic acid, .alpha.-aminodiphenylacetic acid, .alpha.,.alpha.-diphenylglycine, benzeneacetic acid, .alpha.-amino-.alpha.-phenyl, aminodiphenylacetic acid, phenytoin impurity c
PubChem CID 18289
IUPAC Name 2-amino-2,2-diphenylacetic acid
SMILES NC(C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 244.0°C to 247.0°C
Color White
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2NC(C6H5)2CO2H
Quantity 25 g
Beilstein 14,I,625
Formula Weight 227.26
Percent Purity 98%
Physical Form Powder
Chemical Name or Material α,α-Diphenylglycine
Show More Show Less
EINECSNumber 221-299-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.