Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyliodonium Chloride 98.0+%, TCI America™
SDP

Catalog No. D235625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D235625G 25 g
D23565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D235625G Supplier TCI America Supplier No. D235625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1483-72-3
Molecular Formula C12H10ClI
Molecular Weight (g/mol) 316.57
MDL Number MFCD00011909
InChI Key RSJLWBUYLGJOBD-UHFFFAOYSA-M
Synonym diphenyliodonium chloride, diphenyliodanium chloride, iodonium, diphenyl-, chloride, diphenyliodoniumchloride, c12h10i.cl, iodonium, diphenyl-, chloride 1:1, acmc-1bxx9
PubChem CID 73870
IUPAC Name diphenyliodanium chloride
SMILES [Cl-].[I+](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 316.57
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyliodonium Chloride
RTECSNumber NN6651666
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.