Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyliodonium Trifluoromethanesulfonate 98.0+%, TCI America™
SDP

Catalog No. D225325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D225325G 25 g
D22531G 1 g
D22535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D225325G Supplier TCI America Supplier No. D225325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 66003-76-7
Molecular Formula C13H10F3IO3S
Molecular Weight (g/mol) 430.18
MDL Number MFCD00191356
InChI Key SBQIJPBUMNWUKN-UHFFFAOYSA-M
Synonym diphenyliodonium trifluoromethanesulfonate, diphenyliodanium triflate, diphenyliodonium triflate, acmc-209nt6, diphenyliodonium trifluoromethansulfonat, diphenyliodanium; trifluoromethanesulfonate, diphenyliodonium trifluoromethane sulfonate, diphenyliodonium; trifluoromethanesulfonate, diphenyliodanium; tris fluoranyl methanesulfonate
PubChem CID 2737137
IUPAC Name diphenyliodanium;trifluoromethanesulfonate
SMILES C1=CC=C(C=C1)[I+]C2=CC=CC=C2.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Melting Point 178°C
Color White-Yellow
Quantity 25 g
Formula Weight 430.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyliodonium Trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.