Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylphosphinic Acid 98.0+%, TCI America™
SDP

Catalog No. D362725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D362725G 25 g
D36275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D362725G Supplier TCI America Supplier No. D362725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1707-03-5
Molecular Formula C12H11O2P
Molecular Weight (g/mol) 218.19
MDL Number MFCD00002132
InChI Key BEQVQKJCLJBTKZ-UHFFFAOYSA-N
Synonym phosphinic acid, diphenyl, hydroxydiphenylphosphine oxide, diphenylphosphinicacid, diphenyl phosphinic acid, diphenyl-phosphinic acid, acmc-1boli, di phenyl phosphinic acid, p,p-diphenylphosphinic acid, ksc180o2h, phosphinicacid, p,p-diphenyl
PubChem CID 15567
ChEBI CHEBI:37832
IUPAC Name diphenylphosphinic acid
SMILES OP(=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 195°C
Color White-Yellow
Quantity 25 g
Formula Weight 218.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenylphosphinic Acid
RTECSNumber SZ5315000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.