Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylphosphinic Anhydride 98.0+%, TCI America™
SDP

Catalog No. D24841G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24841G 1 g
D24845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24841G Supplier TCI America Supplier No. D24841G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5849-36-5
Molecular Formula C24H20O3P2
Molecular Weight (g/mol) 418.369
MDL Number MFCD01112176
InChI Key XTAYANNRWHXQOL-UHFFFAOYSA-N
PubChem CID 613006
IUPAC Name [diphenylphosphoryloxy(phenyl)phosphoryl]benzene
SMILES C1=CC=C(C=C1)P(=O)(C2=CC=CC=C2)OP(=O)(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 146°C
Color White
Quantity 1 g
Formula Weight 418.37
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenylphosphinic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.