Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylphosphoryl Azide 97.0+%, TCI America™
SDP

Catalog No. D167225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D167225G 25 g
D16725G 5 g
D1672250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D167225G Supplier TCI America Supplier No. D167225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26386-88-9
Molecular Formula C12H10N3O3P
Molecular Weight (g/mol) 275.20
MDL Number MFCD00001987
InChI Key SORGEQQSQGNZFI-UHFFFAOYSA-N
Synonym diphenylphosphoryl azide, diphenyl azidophosphate, diphenylphosphonic azide, diphenyl phosphoryl azide, diphenyl phosphorazidate, phosphorazidic acid, diphenyl ester, azido phenoxy phosphoryl oxy benzene, dppa polymer-bound, diphenylphosphorazidate, unii-gxm91165av
PubChem CID 123414
IUPAC Name {[azido(phenoxy)phosphoryl]oxy}benzene
SMILES [N-]=[N+]=NP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Color Yellow
Quantity 25 g
UN Number 3278
Formula Weight 275.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diphenylphosphoryl Azide
EINECSNumber (3)-3359
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.