Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetone HPLC Plus for HPLC
Supplier: Sigma Aldrich Fine Chemicals Biosciences 6505014X4L
Acetone is a ketone. Mixtures of TiO2 (P25) with rare earth oxides (La2O3 or Y2O3) calcined at 700C or 650C has been reported to efficiently photocatalyze oxidation of acetone. Its aldol condensation in vapor-phase has been investigated over MgO promoted with alkali (Li Na K and Cs) or alkaline earth (Ca Sr and Ba) metal ions. It undergoes Aldol condensation in the presence of Mg-Al layered double hydroxides (LDH) as catalysts and Cl- and/or CO32- as compensating anions to afford diacetone alcohol and mesityl oxide. Enantioselective Aldol condensation of acetone with various isatins in the presence of dipeptides (catalyst) has been described.