Learn More
Creative Biomart Diphtheria Toxin Mutant (CRM197)
Supplier:  Creative Biomart THP0239
Encompass_Preferred
CRM197 is a non-toxic mutant of diphtheria toxin1,2 having a single amino acid substitution of glutamic acid for glycine.
Catalog No. 50-208-4290
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.