Learn More
TARGETMOL CHEMICALS INC DIPHYLLIN 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TN1103
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Diphyllin could be characterized as a new V-ATPase inhibitor in treating gastric cancer and inhibiting the phosphorylation of LRP6 in Wnt/(beta)-catenin signaling. purity: 99%
Catalog No. 50-257-8896
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.