Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipropylamine 99.0+%, TCI America™
SDP

Catalog No. D0930100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0930100ML 100 mL
D0930500ML 500 mL
D093025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0930100ML Supplier TCI America Supplier No. D0930100ML

Chemical Identifiers

CAS 142-84-7
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00009362
InChI Key WEHWNAOGRSTTBQ-UHFFFAOYSA-N
Synonym dipropylamine, di-n-propylamine, 1-propanamine, n-propyl, n-dipropylamine, n-propyl-1-propanamine, rcra waste number u110, dipropyl amine, rcra waste no. u110, unii-60p318iiry, ccris 4805
PubChem CID 8902
IUPAC Name dipropylamine
SMILES CCCNCCC

Specifications

Melting Point -63°C
Color Yellow
Boiling Point 108°C
Quantity 100 mL
UN Number 2383
Formula Weight 101.19
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Dipropylamine
EINECSNumber (2)-3086
RTECSNumber JL9200000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.