Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipropylene Glycol Methyl Ether Acetate, 98%, Spectrum™

Click to view available options
Quantity:
250 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Steel Can
88917-22-0
88917-22-0
Packaging Amber Glass Bottle
Quantity 250 mL
Acidity 0.0005
Water 0.0005
CAS Max % 1
Percent Purity 0.98
Grade Reagent

CAUTION: For use by qualified personnel for research and development or other authorized use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.