Learn More
Abcam Lithocholic acid, endogenous MDM2 and MDM4 selective inhibitor, 5G
Supplier: Abcam AB1455985G
MW 376.6 Da, Purity >97%. Endogenous MDM2 and MDM4 selective inhibitor. Detergent. Solubilizes dietary fats for absorption. Increases p53 and cyclin D1 promoter activity and endogenous p53 and cyclin D1 expression and apoptosis. Shows cancer cell selective antiproliferative effects. Shows cholagogue and choleretic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.