Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α,α-Dipyridyl, 99.5%, MP Biomedicals™

Catalog No. ICN10161401 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10161401 1 g
ICN10161425 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10161401 Supplier MP Biomedicals, Inc Supplier No. 0210161401
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 366-18-7 | C10H8N2 | 156.19 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 366-18-7
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD00006212
InChI Key ROFVEXUMMXZLPA-UHFFFAOYSA-N
Synonym 2,2'-bipyridine, 2,2'-dipyridyl, 2,2'-bipyridyl, bipyridine, 2,2'-dipyridine, 2-2-pyridyl pyridine, 2,2'-bipyridin, alpha,alpha'-dipyridyl, alpha,alpha'-bipyridyl, 2,2-bipyridyl
PubChem CID 1474
ChEBI CHEBI:30351
IUPAC Name 2,2'-bipyridine
SMILES C1=CC=C(N=C1)C1=CC=CC=N1

Specifications

Color White
Assay Percent Range 99.5%
Quantity 1 g
Formula Weight 156.19
Physical Form Powder
Chemical Name or Material α, α-Dipyridyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.