Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α,α'-Dipyridyl (Crystalline/Certified), Fisher Chemical

Catalog No. D955 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
D95-5 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D95-5

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 366-18-7
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD00006212
InChI Key ROFVEXUMMXZLPA-UHFFFAOYSA-N
Synonym 2,2'-bipyridine, 2,2'-dipyridyl, 2,2'-bipyridyl, bipyridine, 2,2'-dipyridine, 2-2-pyridyl pyridine, 2,2'-bipyridin, alpha,alpha'-dipyridyl, alpha,alpha'-bipyridyl, 2,2-bipyridyl
PubChem CID 1474
ChEBI CHEBI:30351
IUPAC Name 2,2'-bipyridine
SMILES C1=CC=C(N=C1)C1=CC=CC=N1

Specifications

Alpha Vector Dipyridyl
Melting Point 69°C
Color Cream
Ignition Residue 0.2% max.
Packaging Glass Bottle
Linear Formula N:CHCH:CHCH:CC:CHCH:CHCH:N
Quantity 5 g
Sensitivity Pass Test
Solubility Information Pass Test (in ethanol)
Formula Weight 156.19
Grade Certified
Physical Form Solid
Chemical Name or Material α, α«-Dipyridyl
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.