Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIRAS2 Monoclonal Antibody (OTI10A6), TrueMAB™, OriGene
SDP

Catalog No. 501682773
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2773 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2773 Supplier OriGene Supplier No. TA809401
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DIRAS2 belongs to a distinct branch of the functionally diverse Ras superfamily of monomeric GTPases.

Specifications

Antigen DIRAS2
Applications Western Blot
Classification Monoclonal
Clone OTI10A6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene DIRAS2
Gene Accession No. Q96HU8
Gene Alias Di-Ras2
Gene Symbols DIRAS2
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-196 of human DIRAS2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54769
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.