Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DIRAS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103870
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5103870 100 μL
1 options

Catalog No. PIPA5103870

Supplier: Invitrogen™ PA5103870

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DIRAS2.

DIRAS2 belongs to a distinct branch of the functionally diverse Ras superfamily of monomeric GTPases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIRAS2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DIRAS2
Gene Accession No. Q5PR73, Q96HU8
Gene Alias 2900052J15Rik; AI414999; DIRAS family GTP binding RAS like 2; DIRAS family GTPase 2; DIRAS family, GTP-binding RAS-like 2; DIRAS2; Di-Ras2; Distinct subgroup of the Ras family member 2; GTP-binding protein Di-Ras2
Gene Symbols DIRAS2
Host Species Rabbit
Immunogen A synthesized peptide derived from human DIRAS2(Accession Q96HU8), corresponding to amino acid residues F162-M199.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291006, 54769, 68203
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.