Description
• A latex-enhanced immunoturbidimetric assay for the automated determination of HbA1c
• For use with routine chemistry analyzers
• Calibrators referenced to NGSP value (assigned HbA1c frozen whole blood)
• Normal and elevated controls are available
• Reagents are liquid stable for cost-effective use of the entire kit
Specifications
Specifications
| Type | HbA1c Control |
| For Use With (Application) | Automated HbA1c Determination |
| Quantity | 4 x 0.5 mL |