Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Direct-Q™ 3 Tap to Pure and Ultrapure Water Purification System
GSA_VA

Catalog No. ZRQSVP300 Shop All MilliporeSigma Products
Change view
Click to view available options
Product Type:
Pure Water Purification System
Ultrapure Water Purification System
Model:
Direct-Q 3UV
Direct-Q 3UV, Without Remote
Product Water Type:
Type 1
Type 3
Electrical Requirements:
100/240 V 50/60 Hz
120 V, 60 Hz
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Model Product Water Type Electrical Requirements
ZRQSVP300 Pure Water Purification System Direct-Q 3UV, Without Remote Type 3 100/240 V 50/60 Hz
ZRQSVP3US Ultrapure Water Purification System Direct-Q 3UV Type 1 120 V, 60 Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ZRQSVP300 Supplier MilliporeSigma™ Supplier No. ZRQSVP300

Requires

Millipore systems always require accessories to make the system operational.

Provide Type I and Type III water from potable tap water with the Direct-Q™ 3 Tap to Pure and Ultrapure Water Purification System.

For laboratories that do not have easy access to pretreated water and need between 1 and 10L of ultrapure water per day.

Key Features

  • Water quality parameters (resistivity or conductivity, temperature) are clearly indicated on bright color display
  • Maintenance is limited to infrequent and easy replacement of required SmartPak DQ3 cartridge (SPR00SIA1US, available separately)
  • Volumetric dispensing function regulates water delivery for volumes up to 2.62 gal. (9.75L) in 0.12 gal. (0.25L) increments Systems connect to potable tap water feed with 1/2 NPTF connector and inlet strainer, and 8mm O.D. tubing (supplied)

 

Direct-Q 3

  • Produces up to >0.1 gal./min. (>0.5L/min.) Type 1 water on demand at 25°C and 30ppb Total Organic Carbon (TOC) directly from potable tap water
  • Product water is suitable for preparing buffers for liquid chromatography separations and biochemical experiments, also for use with such analytical techniques as spectrophotometry and spectroscopy
Direct-Q 3UV
  • Reduces bacteria and TOC to 10ppb via a dual wavelength UV lamp
  • Product water is suitable for applications requiring very low organic contaminants levels such as HPLC, GC, ILC and TOC analyses

 

 

  • Type 1 water for HPLC mobile phase preparation, sample dilution, buffer and cell culture media preparation, preparation of chemical solutions used with titrators, spectrophotometers and electrophoresis systems, preparation of culture media and reagents for molecular biology applications
  • Type 3 water for non-critical applications such as glassware washing or rinsing

 

Certifications

UL listed

†When used with Millipak™ Express™ 20 final filter.

‡When used with BioPak Disposable Ultrafilter.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Bacterial retention >99%
Certifications/Compliance UL, cUL, CE, ISO 9001, ISO 14001
Contaminant Rejection (Microbiological) >99%
Contaminant Rejection (Organic) >99%
Contaminant Rejection (Particulate) >96%
Description Without Remote
Bacterial Content <0.1 CFU/mL
Feedwater Source Potable Tap
Flow Rate 3
Product Water Type Type 3
Resistivity 18.2
Total Organic Carbon <5
For Use With (Application) HPLC, GC, ILC and TOC analyses
Model Direct-Q 3UV, Without Remote
Particle Size >0.22μm, <1 Particulate/mL
Width (English) 11.4 in.
Width (Metric) 29 cm
Height (English) 21.3 in.
Height (Metric) 54 cm
Depth (English) 15 in.
Depth (Metric) 38 cm
Weight (English) 19 lb.
Weight (Metric) 8.6 kg
Electrical Requirements 100/240 V 50/60 Hz
Voltage 100/240 V
Frequency 50/60 Hz
Daily Output 1 to 10 L
Endotoxin Concentration <0.001
Product Type Pure Water Purification System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.