Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Disarticulated Human Rib Bones
SCI_ED

Catalog No. S27454 Shop All Eisco Products
Change view
Click to view available options
Includes:
12 Bones (Half Set)
24 Bones (Full Set)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S27454 24 Bones (Full Set)
S27455 12 Bones (Half Set)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27454 Supplier Eisco™ Supplier No. AMCH1042AS
Add to Cart
Add to Cart

Anatomically correct disarticulated human rib bones are replicas but provide an incredible amount of realism and detail for examining the human skeleton

  • Anatomically correct bones
  • Realistic details, textures, and bony landmarks
  • Ideal for educational and medical demonstrations
  • Use for individual study or as replacement for missing/damaged pieces to a full skeletal model set
  • Made of durable and sturdy PVC plastic
  • Natural size and color
  • Bones are marked for easy demonstration
  • Washable

Specifications

Color Natural
For Use With (Application) Examining The Human Skeleton
Includes 24 Bones (Full Set)
Material PVC
Product Type Anatomical Model, Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.