Learn More
AdipoGen DiSC33
Supplier: AdipoGen CDXD0007M500
Chemical. CAS 53336-12-2. Formula C23H25IN2S2. MW 520.49. Synthetic. Carbocyanine dyes, particularly thiacyanines such as DiSC33 can inhibit respiration and may therefore be relatively cytotoxic. 3,3-dipropyl-thiadicarbocyanine iodide is sensitive to membrane potential, fluorescence response to depolarization depends on the staining concentration and detection method. Selected applications are -Calcium channels and other ion transport systems -Mitochondrial activity -Neurons and brain tissue -Membrane potential in intact yeast cells
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.