Learn More
Discharge Electrode
SDPSCI_ED
Catalog No. S65147 Shop All Science First Products
Change view
Click to view available options
Discharge Electrode
Catalog No.
S65147 Discharge Electrode
1 options
Catalog No. S65147 Supplier Science First Supplier No. N122
Watch how this sphere receives the intense arc discharge from the Van de Graaff generator.
  • When grounded, this polished anodized aluminum sphere is used to receive the intense arc discharges from the Van de Graaff generator dome
  • Grounding lead is 90cm long
  • Banana plug at one end, alligator clip at the other
  • Sphere is 10cm in diameter; handle is 40cm long
Specifications
For Use With (Application) Used to receive the intense arc discharges from the Van de Graaff generator dome
For Use With (Equipment) Van de Graaff generator
Includes Acrylic rod assembled to 100mm dia. polished anodized Aluminum sphere, separate 90cm L grounding Lead wire with banana plug and alligator clip
Product Type Discharge Electrode
WARNING: Cancer - www.P65Warnings.ca.gov