Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Discharge Tubes for Dispensette™ S Bottletop Dispensers

Catalog No. 13882270 Shop All BrandTech Scientific Products
Change view
Click to view available options
Includes:
Recirculation valve
Without recirculation valve
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
13-882-270 Without recirculation valve
13-882-271 Recirculation valve
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-882-270 Supplier BRANDTECH™ BRAND™ Supplier No. 708012
Add to Cart
Add to Cart

For use with Dispensette S Bottletop Dispensers.

  • Fine tip
  • Volume: 5 and 10mL
  • Available with or without recirculation valve
TRUSTED_SUSTAINABILITY

Specifications

Product Type Discharge Tube
Volume (Metric) 5, 10 mL
For Use With (Equipment) Dispensette S Bottletop Dispensers
Includes Without recirculation valve
Tip Type Fine tip
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.