Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Discovery Education™ Health CME: 34-Pack DVD

Offers an entirely new way to learn about latest advances in medicine.
Supplier: Aldon Corporation VNIX0246CS/4
Description
Series provides credible information addressing up-to-date scientific findings and current standards of care for a variety of health topics
- Real-life patient stories
- w/Vivid graphics and animations
- Illustrate relevant health issues facing society
- Format: DVD
- Grade Level: 9 to 12
- Runtime: 1781 min.
- Copyright: 2010
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction