Learn More
Ibidi USA -Dish 35 mm, high ESS, 30/box
SDP
Supplier:  Ibidi USA 81199
Encompass
A 35mm imaging dish with an elastically supported surface (ESS) used for growing cells under "in vivo-near" conditions. High resolution microscopy on soft cell culture surface based on the standard µ-Dish 35mm, high. Pcs./Box: 30 (individually packed)Elastic Surface ESS: ESS 28 kPaSurface Modification: ESS: uncoated, hydrophobic, sterilized
Catalog No. NC1903384
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.