Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disodium 1,4-Butanedisulfonate 98.0+%, TCI America™
SDP

Catalog No. B067610G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B067610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B067610G Supplier TCI America Supplier No. B067610G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 36589-61-4
Molecular Formula C4H8Na2O6S2
Molecular Weight (g/mol) 262.20
MDL Number MFCD00059839
InChI Key LEUIUWYZAHKPSE-UHFFFAOYSA-L
Synonym 1,4-Butanedisulfonic Acid Disodium Salt
PubChem CID 119021
IUPAC Name disodium butane-1,4-disulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)CCCCS([O-])(=O)=O

Specifications

Color White
Quantity 10 g
Formula Weight 262.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 1,4-Butanedisulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.