Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disodium 3,3'-Dithiobis(1-propanesulfonate) 97.0+%, TCI America™
SDP

Catalog No. D5159100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5159100G 100 g
D515925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5159100G Supplier TCI America Supplier No. D5159100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27206-35-5
Molecular Formula C6H12Na2O6S4
Molecular Weight (g/mol) 354.376
MDL Number MFCD01104840
InChI Key WIYCQLLGDNXIBA-UHFFFAOYSA-L
Synonym 3,3′-Dithiobis(1-propanesulfonic Acid) Disodium Salt
PubChem CID 117948
IUPAC Name disodium;3-(3-sulfonatopropyldisulfanyl)propane-1-sulfonate
SMILES C(CSSCCCS(=O)(=O)[O-])CS(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color White
Quantity 100 g
Formula Weight 354.38
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 3,3′-Dithiobis(1-propanesulfonate)
EINECSNumber (2)-1602
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.