Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disodium 4,4'-Bis(2-sulfonatostyryl)biphenyl 97.0+%, TCI America™
SDP

Catalog No. D3251100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3251100G 100 g
D325125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3251100G Supplier TCI America Supplier No. D3251100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 27344-41-8
Molecular Formula C28H20Na2O6S2
Molecular Weight (g/mol) 562.56
MDL Number MFCD00318874,MFCD00318874
InChI Key PMPJQLCPEQFEJW-GNTLFSRWSA-L
PubChem CID 87075690
IUPAC Name disodium 2-[(1Z)-2-{4'-[(1Z)-2-(2-sulfonatophenyl)ethenyl]-[1,1'-biphenyl]-4-yl}ethenyl]benzene-1-sulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC=C1\C=C/C1=CC=C(C=C1)C1=CC=C(\C=C/C2=CC=CC=C2S([O-])(=O)=O)C=C1

Specifications

Color Yellow
Quantity 100 g
Synonym 4,4′-Bis(2-sulfonatostyryl)biphenyl Disodium Salt
Formula Weight 562.56
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 4,4′-Bis(2-sulfonatostyryl)biphenyl
EINECSNumber (5)-4880
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.