Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disodium 4,4'-Dinitrostilbene-2,2'-disulfonate 98.0+%, TCI America™
SDP

Catalog No. D0853100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0853100G 100 g
D085325G 25 g
D0853500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0853100G Supplier TCI America Supplier No. D0853100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3709-43-1
Molecular Formula C14H8N2Na2O10S2
Molecular Weight (g/mol) 474.32
MDL Number MFCD00024929
InChI Key SDCDTWFAOKXZHD-SEPHDYHBSA-L
PubChem CID 21116162
IUPAC Name disodium 5-nitro-2-[(1E)-2-(4-nitro-2-sulfonatophenyl)ethenyl]benzene-1-sulfonate
SMILES [Na+].[Na+].[O-][N+](=O)C1=CC=C(\C=C\C2=CC=C(C=C2S([O-])(=O)=O)[N+]([O-])=O)C(=C1)S([O-])(=O)=O

Specifications

Quantity 100 g
Synonym 4,4′-Dinitrostilbene-2,2′-disulfonic Acid Disodium Salt
Formula Weight 474.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 4,4′-Dinitrostilbene-2,2′-disulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.