Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Disopyramide Monoclonal Antibody (DS11-420.3)
GREENER_CHOICE

Catalog No. 01671948
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-948 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-948 Supplier Invitrogen™ Supplier No. MA110718
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-10718 detects Disopyramide. MA1-10718 has been successfully used in ELISA applications. The MA1-10718 immunogen is Disopyramide, conjugated to KLH.

Disopyramide is a class I anti-arrhythmic agent (one that interferes directly with the depolarization of the cardiac membrane and thus serves as a membrane-stabilizing agent) with a depressant action on the heart similar to that of guanidine. It also possesses some anticholinergic and local anesthetic properties. [National Center for Biotechnology Information. PubChem Compound Database; CID=3114].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Disopyramide
Applications ELISA
Classification Monoclonal
Clone DS11-420.3
Concentration ~1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias Dicorantil; Disopyramide; Norpace; Ritmodan; Rythmodan
Host Species Mouse
Immunogen Disopyramide, conjugated to KLH.
Purification Method Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.