Learn More
Invitrogen™ Disopyramide Monoclonal Antibody (DS11-433.5)

Description
MA1-10719 detects Disopyramide. MA1-10719 has been successfully used in ELISA applications. The MA1-10719 immunogen is Disopyramide, conjugated to KLH.
Specifications
Specifications
| Antigen | Disopyramide |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | DS11-433.5 |
| Concentration | ∼1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene Alias | Dicorantil; Disopyramide; Norpace; Ritmodan; Rythmodan |
| Host Species | Mouse |
| Immunogen | Disopyramide, conjugated to KLH. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.