Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DISP2 Monoclonal Antibody (758204)
GREENER_CHOICE

Catalog No. MA561953
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA561953 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA561953 Supplier Invitrogen™ Supplier No. MA561953
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Reconstitute at 0.5 mg/mL in sterile PBS.

DISP2, also known as dispatched homolog 2, is a protein that plays a critical role in the regulation of the hedgehog signaling pathway. It is involved in the transport and release of hedgehog ligands, which are important for cellular differentiation and tissue development. Mutations in the DISP2 gene have been associated with various developmental disorders, including holoprosencephaly and cleft lip and palate. DISP2 is also being studied as a potential therapeutic target in cancer, as the hedgehog pathway is frequently dysregulated in various types of tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DISP2
Applications Western Blot
Classification Monoclonal
Clone 758204
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene DISP2
Gene Accession No. A7MBM2
Gene Alias AI840597; B230210L08Rik; C15orf36; DISP2; dispatched B; dispatched homolog 2; dispatched homolog 2 (Drosophila); dispatched RND transporter family member 2; DISPB; HsT16908; KIAA1742; LINC00594; mKIAA1742; Protein dispatched homolog 2
Gene Symbols DISP2
Host Species Mouse
Immunogen E. coli-derived recombinant human DISP2, Arg192-Lys477.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85455
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.