Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Dispase (B. polymyxa)

Catalog No. 89942016 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-942-016 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-942-016 Supplier Abnova™ Supplier No. P5318
Add to Cart
Add to Cart

Used for Enzyme

#REF!

Specifications

Accession Number P29148
For Use With (Application) Enzyme Activity
Molecular Weight (g/mol) 36kDa
Name Dispase (B. polymyxa)
Preparation Method Native protein purified from Bacillus polymyxa
Quantity 1 g
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name DISPASE
Assays Neutral Protease (Dispase) Assay; Tissue disaggregation and subcultivation; Prevention of unwanted cell clumping; Preparation of cells for culture; Separation of intact epidermis from dermis and intact epithelial sheet in culture from the substratum; Harvest and transfer of normal, diploid cells and cell lines; Gentle and intact detachment of epidermal cells.
Biological Activity ≥0.1μ/mg dry weight. One Unit releases Folin positive amino acids equivalent to 1 umole tyrosine per minute from casein at 37°C, pH 7.5.
Species Bacteria
Protein Tag None
Expression System Native protein purified from Bacillus polymyxa
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.