Learn More
JDM SCIENTIFIC LLC Dispenser for adhesive tapes
Supplier:  JDM SCIENTIFIC LLC 01172
Encompass_Preferred
The tape or labeling tape dispenser is designed to hold up to 6 rolls with a core hole diameter of approximately 25 mm (1 inch) or approximately 76 mm (3 inches) with a total width of up to 160 mm. It is made of powder-coated steel, polyamide 12 and stainless steel. The serrated cutting device for easy cutting of the adhesive tape or labeling tape facilitates one-handed handling in the laboratory and a clean cut. The dispenser for adhesive tape or labeling tape is easy to clean.Two cores are included to accommodate rolls with a core hole diameter of approx. 25 mm (1 inch) or approx. 76 mm (3 inches).
Catalog No. 50-255-8537
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.