Learn More
Shamrock Supply Company Inc Lakeland Disposable Coverall, 05414, 3XL, Yellow, DuPont Tychem QC,
Supplier:  Shamrock Supply Company Inc C1S414Y3X
Encompass
Lakeland Disposable Coverall, Series: 05414, 3XL, Yellow, DuPont Tychem QC, 56 - 58 in Chest, 29 in Inseam Length, 2.1 oz/sq-yd Fabric Weight, Elastic Cuff, Serged Seam, Zipper Closure 25/case
Catalog No. NC3360748
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.