Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Disposable Aluminum Moisture Balance Pans

Catalog No. 01001766
Change view
Click to view available options
Material:
Aluminum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
01-001-766 Aluminum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-001-766 Supplier Qorpak™ Supplier No. 234794
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Made from oil free aluminum to provide even temperatures across each pan

  • With crimped sides and flat smooth bottom
  • Diameter x D: 5 x 0.375 in. (127 x 9.52 mm)
  • Compatible with most moisture balances
  • For use with liquid and solid samples including weighing, drying and moisture determination

Specifications

CAS 67-56-1, 67812-42-4
Model 234794
Material Aluminum
Linear Formula C26H36N2O4 · HCl
MDL Number MFCD00078605
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Height (English) 5 in.
Concentration 1.0 mg/mL in methanol (as free base)
Type Aluminum Pan
Formula Weight 477.04
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.