Learn More
Grainger ECOBAILER Disposable Bailer, Groundwater
Supplier:  Grainger 9MFD0
Encompass_Preferred
The ECOBAILER Disposable Bailer from Grainger is a single-use bailer designed for groundwater sampling. It features a polypropylene construction with a check valve in the bottom to prevent backflow. The bailer has a capacity of 1 liter and a working depth of 100 feet. It can be used to collect groundwater samples for testing. The disposable design eliminates cross-contamination.
Catalog No. NC2676369
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.