Learn More
Ted Pella Inc Disposable Base Molds, 30 x 24 x 5mm, 500/box
SDP
Supplier:  Ted Pella Inc 271474
Encompass
Disposable base molds offer ease and convenience of operation. They are inexpensive enough to be discarded after use, yet strong enough to be reused. Made of PVC, they offer excellent thermal exchange. They have a smooth interior finish and rounded corners to facilitate their removal.
Catalog No. NC0764543
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.