Learn More
StatLab DISPOSABLE BLADE REMOVER
Supplier: StatLab BLR00201P
This blade remover can hold 150-300 blades. Works on any blade from #10-22, but will not work on double edged blades.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.