Learn More
Andwin Scientific DISPOSABLE EMBED.MOULDS 266/CS
SDP
Supplier:  Andwin Scientific 15160215
Encompass
The Peel-A-Way Disposable Embedding Molds are paraffin molds that are ideal for plastic or paraffin embedding. This product removes the necessity of elease agent, additional parts, and block trimming. The molds have a unique design that allows the "Peel-A-Way" from the solidified embedded block. This is sold in a case of 266.
Catalog No. NC9613045