Learn More
New Pig Disposable Face Shield
SDP
Catalog No. 17987514 Shop All New Pig Corporation Products
Change view
Click to view available options
No. per Pack:
25/Pk.
50/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
17-987-514 50/Pk.
17-987-513 25/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-987-514 Supplier New Pig Supplier No. GLS21850P
Add to Cart
Add to Cart
Lightweight, fog-free protection you can trust.
  • Provides full-face protection against droplets and splashes
  • Anti-fog interior coating ensures visibility during prolonged use
  • Foam padding enhances comfort and breathability
  • Elastic band offers a universal fit
  • Write names or roles on the white band
Specifications
Lens Coating Anti-fog
Lens Tint Clear
Product Type Faceshield
Headband Material Elastic, Foam
No. per Pack 50/Pk.
Material PET
Disposable Single-use
Lens Material PET
Frame Material Plastic
UV Protection None
Autoclavable Not Autoclavable