Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Disposable Glass Test Tubes
SDPSCI_ED

Catalog No. S72381 Shop All LaMotte Company Products
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S72381 10 mL
S72379 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S72381 Supplier LaMotte™ Supplier No. 0843
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Accessory for test kits

  • Material: glass
  • With cap

Specifications

Capacity (Metric) 10 mL
Material Glass
For Use With For test kits
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.