Learn More
Fisherbrand™ Disposable Inoculating Loops
Catalog No. p-7236027 Shop All Fisherbrand Products
Change view
Click to view available options
Volume:
1 μL
10 μL
Sterility:
Non-sterile
Sterile
Quantity:
Cs of 1000 (40 Pks. of 25)
Cs of 1000 (50 Pks. of 20)
Cs of 125 (5 Pks. of 25/ Individually Packed)
Cs of 250 (5 Pks. of 50/ Individually Packed)
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume Sterility Quantity
01-189-162 1 μL Sterile Cs of 1000 (50 Pks. of 20)
01-189-163 1 μL Non-sterile Cs of 1000 (50 Pks. of 20)
01-189-164 1 μL Sterile Cs of 1000 (40 Pks. of 25)
01-189-165 1 μL Sterile Cs of 250 (5 Pks. of 50/ Individually Packed)
01-189-166 10 μL Sterile Cs of 1000 (50 Pks. of 20)
01-189-167 10 μL Non-sterile Cs of 1000 (50 Pks. of 20)
01-189-168 10 μL Sterile Cs of 1000 (40 Pks. of 25)
01-189-169 10 μL Sterile Cs of 125 (5 Pks. of 25/ Individually Packed)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 01-189-162
These Disposable Inoculating Loops are smooth and flexible to facilitate smooth, uniform streaking without damaging gel surfaces. Sterile and non-sterile options available.
  • Ideal for anaerobic microbiology applications where deep agar inoculation is required
  • Use in situations where flaming is not practical, such as under hoods and in anaerobic chamber
  • High-impact polystyrene loops are smooth and flexible to facilitate smooth, uniform streaking without damaging the gel surface
  • Rigid enough to penetrate the media for subsurface inoculation
  • Disposable loops do not need flaming, thus help to eliminate the risk of infection due to aerosol formation of pathogenic substances
  • Can be used under hoods without danger
  • Color-coded for ease of size identification
Specifications
Disposable Single-use
Volume 1 μL
Material Polystyrene
Sterility Sterile
Color White
Quantity Cs of 1000 (50 Pks. of 20)
Autoclavable Not Autoclavable