Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tians Disposable Polypropylene Coveralls

Catalog No. 191302073G Shop All Tians International Products
Change view
Click to view available options
Size:
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-2073G 4X-Large
19-130-2073D X-Large
19-130-2073E 2X-Large
19-130-2073F 3X-Large
19-130-2073H 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-130-2073G Supplier Tians Supplier No. 2108824XL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Low lint coverall for use in ISO cleanroom class 7 critical environments and many industrial applications. Nonwoven fabric is cloth-like and cool to wear.

  • Economical white polypropylene coverall is ideal low-cost alternative for general protection in nonhazardous environments
  • Cool, comfortable, and lightweight
  • Zipper front, elastic wrist and ankles, and folded collar

Specifications

Product Type Cleanroom Coveralls
Size 4X-Large
Material Polypropylene
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Closure Type Front Zipper
Sterility Non-sterile
For Use With (Application) Controlled Environments
Cleanroom Class Class 10,000 (ISO 7)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.